Learn More
1,1-Dichloroethane, SPEX CertiPrep™
Catalog Number 12939174
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
1,1-Dichloroethane75-34-31000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
CAS 75-34-3, 67-56-1
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Molecular Formula C2H4Cl2
InChI Key SCYULBFZEHDVBN-UHFFFAOYSA-N
SMILES CC(Cl)Cl
IUPAC Name 1,1-dichloroethane
Molecular Weight (g/mol) 98.954
PubChem CID 6365
ChEBI CHEBI:81599
Packaging 2mL amber prescored glass ampule
Solvent or Matrix Methanol-P&T
Quantity 1 mL
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Chemical Name or Material 1,1-Dichloroethane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol
Product intended for laboratory use only