missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Diethyl-2,2'-carbocyanine iodide, 96%

Product Code. p-7030709
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15454137

Brand: Thermo Scientific Alfa Aesar H31540.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 605-91-4 | C25H25IN2 | 480.39 g/mol

The fluorescence decay time of pinacyanol (1,1?-diethyl-2,2?-carbocyanine) iodide has been measured in the temperature range 95-225 K.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The fluorescence decay time of pinacyanol (1,1′-diethyl-2,2′-carbocyanine) iodide has been measured in the temperature range 95-225 K.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-91-4
Molecular Formula C25H25IN2
Molecular Weight (g/mol) 480.39
MDL Number MFCD00011975
InChI Key QWYZFXLSWMXLDM-UHFFFAOYSA-M
Synonym carbocyanine, pinacyanole, pinacyanol iodide, sensitol red, pinacyanol, eastman kodak 622, quinaldine blue, dsstox_cid_26576, dsstox_rid_81735, dsstox_gsid_46576
PubChem CID 5709754
ChEBI CHEBI:52218
SMILES [I-].CCN1C(=CC=CC2=CC=C3C=CC=CC3=[N+]2CC)C=CC2=CC=CC=C12

Specifications

Melting Point >250°C
Quantity 1 g
UN Number UN2811
IUPAC Name (2Z)-1-ethyl-2-[(E)-3-(1-ethylquinolin-1-ium-2-yl)prop-2-enylidene]quinoline;iodide
Formula Weight 480.38
Percent Purity 96%
Chemical Name or Material 1,1'-Diethyl-2,2'-carbocyanine iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.