missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylbiguanide hydrochloride, 97%

Product Code. 10593658 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10593658 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10593658 Supplier Thermo Scientific Acros Supplier No. 429720050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1115-70-4 | C4H12ClN5 | 165.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1115-70-4
CAS Min % 96.0
CAS Max % 100.0
Melting Point 218.0°C to 223.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C4H12ClN5
MDL Number MFCD00012582
Quantity 5 g
Merck Index 15,601
Synonym metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,dmso (heated)
InChI Key OETHQSJEHLVLGH-UHFFFAOYSA-N
SMILES [H+].[Cl-].CN(C)C(=N)N=C(N)N
IUPAC Name 3-(diaminomethylidene)-1,1-dimethylguanidine;hydrochloride
Molecular Weight (g/mol) 165.63
PubChem CID 14219
Formula Weight 165.63
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1, 1-Dimethylbiguanide hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.