missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenylbut-1-ene, Mikromol™

Product Code. 17903616
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17903616 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17903616 Supplier Mikromol Supplier No. MM0760.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,1-Diphenylbut-1-ene
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Diphenylbut-1-ene
CAS 1726-14-3
Analyte or Component Names 1,1-Diphenylbut-1-ene
Shipping Condition Room Temperature
Molecular Formula C16 H16
Certifications/Compliance ISO 9001
Synonym 1,1-Diphenylbut-1-ene
InChI Formula InChI=1S/C16H16/c1-2-9-16(14-10-5-3-6-11-14)15-12-7-4-8-13-15/h3-13H,2H2,1H3
SMILES CCC=C(c1ccccc1)c2ccccc2
IUPAC Name 1-phenylbut-1-enylbenzene
Molecular Weight (g/mol) 208.3
Quantity 100 mg
Formula Weight 208.1252 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.