Learn More
1,1'-Ethylidenebis[L-tryptophan], TRC
Product Code. 30012669
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012669 50 mg 50mg
30003794 100 mg 100mg
30012472 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012669 Supplier Toronto Research Chemicals Supplier No. TRCE91988050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 132685-02-0
Molecular Formula C24 H26 N4 O4
Molecular Weight (g/mol) 434.49
Synonym 3,3'-[Ethylidenebis(1H-indole-1,3-diyl)]bis[(2S)-2-aminopropanoic] Acid, Tryptophan Imp. A (EP), 1,1'-Ethylidenebistryptophan
IUPAC Name (2S)-2-amino-3-[1-[1-[3-[(2S)-2-amino-2-carboxyethyl]indol-1-yl]ethyl]indol-3-yl]propanoic acid
SMILES CC(n1cc(C[C@H](N)C(=O)O)c2ccccc12)n3cc(C[C@H](N)C(=O)O)c4ccccc34
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C24H26N4O4/c1-14(27-12-15(10-19(25)23(29)30)17-6-2-4-8-21(17)27)28-13-16(11-20(26)24(31)32)18-7-3-5-9-22(18)28/h2-9,12-14,19-20H,10-11,25-26H2,1H3,(H,29,30)(H,31,32)/t19-,20-/m0/s1
Formula Weight 434.1954
Percent Purity >95%
Chemical Name or Material 1,1'-Ethylidenebistryptophan
Grade HPLC