Learn More
1-(1-Methyl-3,3-diphenylpropyl)piperidine Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
1-(1-Methyl-3,3-diphenylpropyl)piperidine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 1-(1-Methyl-3,3-diphenylpropyl)piperidine Hydrochloride |
| CAS | 110246-09-8 |
| Analyte or Component Names | 1-(1-Methyl-3,3-diphenylpropyl)piperidine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H27 N . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-(1-Methyl-3,3-diphenylpropyl)piperidine Hydrochloride |
| InChI Formula | InChI=1S/C21H27N.ClH/c1-18(22-15-9-4-10-16-22)17-21(19-11-5-2-6-12-19)20-13-7-3-8-14-20;/h2-3,5-8,11-14,18,21H,4,9-10,15-17H2,1H3;1H |
| SMILES | Cl.CC(CC(c1ccccc1)c2ccccc2)N3CCCCC3 |
| IUPAC Name | 1-(4,4-diphenylbutan-2-yl)piperidine;hydrochloride |
| Show More |