missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals

Catalog Number p-7049904
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5144-89-8
C12H10N2O
198.23
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
21226
1,10-phenanthroline;hydrate
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5144-89-8
C12H10N2O
198.23
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
21226
1,10-phenanthroline;hydrate
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 97°C to 101°C
Quantity 10 g
UN Number UN2811
Beilstein 4008096
Merck Index 14,7214
Solubility Information Soluble in alcohol,acetone; Slightly soluble in waterSoluble in ethanol,acetone and methanol; Slightly soluble in water and benzene.
Formula Weight 198.23 (180.21 Anhydrous)
Grade ACS
Physical Form Crystalline
Chemical Name or Material 1, 10-Phenanthroline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'