missing translation for 'onlineSavingsMsg'
Learn More

[1-[[[(1R)-1-[3-[(E)-2-(7-Chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(1-methylethenyl)phenyl]propyl]sulfanyl]methyl]cyclopropyl]acetic Acid, Mikromol™

Product Code. 30051270
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051270 100 mg 100mg
30051803 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051270 Supplier Mikromol Supplier No. MM0698.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[1-[[[(1R)-1-[3-[(E)-2-(7-Chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(1-methylethenyl)phenyl]propyl]sulfanyl]methyl]cyclopropyl]acetic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Montelukast Styrene
CAS 918972-54-0
Analyte or Component Names Montelukast Styrene
Shipping Condition Room Temperature
Molecular Formula C35 H34 Cl N O2 S
Certifications/Compliance ISO 9001
Synonym Montelukast Sodium Imp. B (EP), Montelukast Imp. B (EP), 1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-methylethenyl)phenyl]propyl]thio]methyl]-cyclopropaneacetic Acid, Montelukast Styrene, Montekulast Sodium Imp. B (EP), Montelukast Sodium Impurity B, Montelukast Impurity B
InChI Formula InChI=1S/C35H34ClNO2S/c1-24(2)31-9-4-3-7-26(31)13-17-33(40-23-35(18-19-35)22-34(38)39)28-8-5-6-25(20-28)10-15-30-16-12-27-11-14-29(36)21-32(27)37-30/h3-12,14-16,20-21,33H,1,13,17-19,22-23H2,2H3,(H,38,39)/b15-10+/t33-/m1/s1
SMILES CC(=C)c1ccccc1CC[C@@H](SCC2(CC(=O)O)CC2)c3cccc(\C=C\c4ccc5ccc(Cl)cc5n4)c3
IUPAC Name 2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-(2-prop-1-en-2-ylphenyl)propyl]sulfanylmethyl]cyclopropyl]acetic acid
Molecular Weight (g/mol) 568.17
Quantity 100 mg
Formula Weight 567.1999 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.