Learn More
1,2,3,4-Tetra-O-acetyl-b-D-glucuronic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2,3,4-Tetra-O-acetyl-beta-D-glucuronic Acid |
| CAS | 62133-77-1 |
| Molecular Formula | C14 H18 O11 |
| Quantity | 250 mg |
| Synonym | 1,2,3,4-Tetra-O-Acetyl-B-D-Glucuronic Acid |
| InChI Formula | InChI=1S/C14H18O11/c1-5(15)21-9-10(22-6(2)16)12(23-7(3)17)14(24-8(4)18)25-11(9)13(19)20/h9-12,14H,1-4H3,(H,19,20)/t9-,10-,11-,12+,14+/m0/s1 |
| SMILES | CC(=O)O[C@@H]1O[C@@H]([C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]1OC(=O)C)C(=O)O |
| IUPAC Name | (2S,3S,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxane-2-carboxylic acid |
| Molecular Weight (g/mol) | 362.29 |
| Formula Weight | 362.0849 |