missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose, 98%

Product Code. p-7043219
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400557 500 mg 500mg
15497609 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400557 Supplier Thermo Scientific Chemicals Supplier No. J66307.ME
Add to basket
This item is not returnable. View return policy

CAS: 13100-46-4 | C14H20O10 | 348.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13100-46-4
Molecular Formula C14H20O10
Molecular Weight (g/mol) 348.30
MDL Number MFCD00063260
InChI Key FEQXFAYSNRWXDW-UHFFFAOYNA-N
Synonym 1,2,3,4-tetra-o-acetyl-beta-d-glucopyranose, beta-d-glucose 1,2,3,4-tetraacetate, 2s,3r,4s,5r,6r-6-hydroxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, 2r,3r,4s,5r,6s-4,5,6-tris acetyloxy-2-hydroxymethyl oxan-3-yl acetate, 1,2,3,4-tetra-o-acetylglucopyranose, beta-d-glucopyranose 1,2,3,4-tetraacetate, 1,2,3,4-tetra-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose, 1,2,3,4-tetraacetate, b-d-glucopyranose,1,2,3,4-tetraacetate
PubChem CID 83136
IUPAC Name [(2R,3R,4S,5R,6S)-4,5,6-triacetyloxy-2-(hydroxymethyl)oxan-3-yl] acetate
SMILES CC(=O)OC1OC(CO)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 126°C to 128°C
Color White
Quantity 500 mg
Solubility Information Soluble in chloroform
Optical Rotation +8° (c=6 in Chloroform)
Formula Weight 348.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.