missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetra-O-acetyl-Beta-D-glucopyranose, TRC

Product Code. 30133166
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133166 25 g 25g
30135691 5 g 5g
30011464 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133166 Supplier Toronto Research Chemicals Supplier No. TRCT28000525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13100-46-4
Formula Weight 348.1056
InChI Formula InChI=1S/C14H20O10/c1-6(16)20-11-10(5-15)24-14(23-9(4)19)13(22-8(3)18)12(11)21-7(2)17/h10-15H,5H2,1-4H3/t10-,11-,12+,13-,14-/m1/s1
IUPAC Name [(2R,3R,4S,5R,6S)-4,5,6-triacetyloxy-2-(hydroxymethyl)oxan-3-yl] acetate
Molecular Formula C14 H20 O10
Molecular Weight (g/mol) 348.3
SMILES CC(=O)O[C@@H]1O[C@H](CO)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]1OC(=O)C
Synonym β-D-Glucopyranose, 1,2,3,4-tetraacetate, Glucopyranose, 1,2,3,4-tetraacetate, β-D- (8CI), 1,2,3,4-Tetra-O-acetyl-β-D-glucopyranose, 1,2,3,4-Tetra-O-acetyl-β-D-glucose, NSC 409250
Chemical Name or Material 1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.