Learn More
1,2,3,4-Tetrahydronaphthalen-1-ol, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 529-33-9 |
| Molecular Formula | C10H12O |
| MDL Number | MFCD00001739 |
| Quantity | 25 g |
| SMILES | OC1CCCC2=C1C=CC=C2 |
| Molecular Weight (g/mol) | 148.20 |
| Percent Purity | 97% |
| Chemical Name or Material | 1,2,3,4-Tetrahydronaphthalen-1-ol |