Learn More
1,2,4,5-Tetramethylbenzene-d14, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Durene-d14 |
| CAS | 15502-50-8 |
| Molecular Formula | C10 D14 |
| Quantity | 0.5 g |
| Synonym | 1,2,4,5-Tetramethylbenzene D14, Benzene-1,4-d2, 2,3,5,6-tetra(methyl-d3)- (9 CI, ACI), 2,3,5,6-Tetra(methyl-d3)benzene-1,4-d2 (ACI), Benzene-1,4-d2, tetra(methyl-d3)- (6 CI), Benzene-p-d2, 2,3,5,6-tetra(methyl-d3)- (8 CI), Durene-d14 |
| InChI Formula | InChI=1 S/C10H14/c1-7-5-9(3)10(4)6-8(7)2/h5-6 H,1-4H3/i1D3,2D3,3D3,4D3,5 D,6 D |
| SMILES | [2 H]c1c(c(c([2 H])c(c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,4-dideuterio-2,3,5,6-tetrakis(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 148.304 |
| Formula Weight | 148.197 g/mol |
| Show More |