missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Triazin-3-amine, 97%, Thermo Scientific™

Product Code. p-4812554
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10352003

Brand: Thermo Scientific Maybridge AC14623DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-99-6
Molecular Formula C3H4N4
Molecular Weight (g/mol) 96.093
InChI Key MJIWQHRXSLOUJN-UHFFFAOYSA-N
Synonym 3-amino-1,2,4-triazine, as-triazine, 3-amino, 3-amino-as-triazine, 1,2,4-triazin-3-ylamine, 1,2,4-triazine-3-ylamine, 1,2,4 triazin-3-ylamine, zlchem 1277, 1,2,4-triazine-3-amine, 3-amino-1,2,4,-triazine
PubChem CID 70715
IUPAC Name 1,2,4-triazin-3-amine
SMILES C1=CN=NC(=N1)N

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 96.09
Chemical Name or Material 1, 2, 4-Triazin-3-amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.