missing translation for 'onlineSavingsMsg'
Learn More

[1,2,4]Triazolo[1,5-a]pyridin-6-ol, TRC

Product Code. 30423836
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30423836 10 mg
30417056 50 mg
30414558 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423836 Supplier Toronto Research Chemicals Supplier No. TRCT13946010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1394969-56-2
Molecular Formula C6H5N3O
Molecular Weight (g/mol) 135.12
IUPAC Name [1,2,4]triazolo[1,5-a]pyridin-6-ol
SMILES Oc1ccc2ncnn2c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C6H5N3O/c10-5-1-2-6-7-4-8-9(6)3-5/h1-4,10H
Formula Weight 135.0433
Chemical Name or Material [1,2,4]Triazolo[1,5-a]pyridin-6-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.