missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(tosyloxy)ethane, TRC

Product Code. 30341916
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30341916 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341916 Supplier Toronto Research Chemicals Supplier No. TRCB419350250MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyleneglycolditosylat
CAS 6315-52-2
Molecular Formula C16 H18 O6 S2
Quantity 250 mg
Synonym Ethylene glycol ditosylat, 1,2-Ethanediol, 1,2-bis(4-methylbenzenesulfonate) (ACI), 1,2-Ethanediol, bis(4-methylbenzenesulfonate) (9CI), Ethylene glycol, di-p-toluenesulfonate (6CI, 8CI), 1,2-Bis(4-methylbenzenesulfonyloxy)ethane, 1,2-Bis(p-toluenesulfonyloxy)ethane, 1,2-Bis(p-tolylsulfonyloxy)ethane, 1,2-Bistosyloxy ethane, 1,2-Di(p-tosyloxy)ethane, 1,2-Di(tosyloxy)ethane, 1,2-Ethanediol ditosylate, 1,2-Ethylene ditosylate, 2-(4-Methylphenyl)sulfonyloxyethyl 4-methylbenzenesulfonate, Di-O-tosylglycol, Ethane-1,2-diyl bis(4-methylbenzenesulfonate), Ethylene 1,2-bis(tosylate), Ethylene di-p-toluenesulfonate, Ethylene glycol 1,2-ditosylate, Ethylene glycol bis(p-toluenesulfonate), Ethylene glycol di-p-tosylate, Ethylene glycol ditoluenesulfonate, Ethylene glycol ditosylate, NSC 21094
InChI Formula InChI=1S/C16H18O6S2/c1-13-3-7-15(8-4-13)23(17,18)21-11-12-22-24(19,20)16-9-5-14(2)6-10-16/h3-10H,11-12H2,1-2H3
SMILES Cc1ccc(cc1)S(=O)(=O)OCCOS(=O)(=O)c2ccc(C)cc2
IUPAC Name 2-(4-methylphenyl)sulfonyloxyethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 370.44
Formula Weight 370.0545
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.