missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one, 95%, Thermo Scientific Chemicals

Catalog Number p-7052552
99.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
84501-67-7
C5H9BrN4O
221.058
MFCD16251259
KQKSCWCKLKLXJS-UHFFFAOYSA-N
1-2-bromoethyl-4-ethyl-1,4-dihydro-5h-tetrazol-5-one, 1-2-bromoethyl-4-ethyltetrazol-5-one, 1-2-bromoethyl-4-ethyl-tetrazol-5-one, 1-2-bromoethyl-4-ethyl-1,2,3,4-tetrazol-5-one, 2-4-ethyl-4,5-dihydro-5-oxo-1h-tetrazol-1-yl ethyl bromide
3019915
1-(2-bromoethyl)-4-ethyltetrazol-5-one
CCN1C(=O)N(N=N1)CCBr
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
84501-67-7
C5H9BrN4O
221.058
MFCD16251259
KQKSCWCKLKLXJS-UHFFFAOYSA-N
1-2-bromoethyl-4-ethyl-1,4-dihydro-5h-tetrazol-5-one, 1-2-bromoethyl-4-ethyltetrazol-5-one, 1-2-bromoethyl-4-ethyl-tetrazol-5-one, 1-2-bromoethyl-4-ethyl-1,2,3,4-tetrazol-5-one, 2-4-ethyl-4,5-dihydro-5-oxo-1h-tetrazol-1-yl ethyl bromide
3019915
1-(2-bromoethyl)-4-ethyltetrazol-5-one
CCN1C(=O)N(N=N1)CCBr

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.777 g/mL
Boiling Point 223°C
Flash Point 89°C (192°F)
Refractive Index 1.507
Quantity 1 g
Formula Weight 221.06
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-(2-Bromoethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'