Learn More
1-(2-Bromophenyl)piperazine, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(2-Bromophenyl)piperazine |
| CAS | 1011-13-8 |
| Molecular Formula | C10 H13 Br N2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C10H13BrN2/c11-9-3-1-2-4-10(9)13-7-5-12-6-8-13/h1-4,12H,5-8H2 |
| SMILES | Brc1ccccc1N2CCNCC2 |
| IUPAC Name | 1-(2-bromophenyl)piperazine |
| Molecular Weight (g/mol) | 241.128 |
| Formula Weight | 240.026 |