missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloro-4-fluorophenyl)ethanol, 95%, Thermo Scientific™

Product Code. 15414607
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414607 5 g 5g
15404607 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414607 Supplier Thermo Scientific Alfa Aesar Supplier No. H31958.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112108-68-6
Molecular Formula C8H8ClFO
Molecular Weight (g/mol) 174.60
MDL Number MFCD04973765
InChI Key UVTJHIWKGYOZFO-UHFFFAOYNA-N
Synonym 1-2-chloro-4-fluorophenyl ethanol, 1-2-chloro-4-fluorophenyl ethan-1-ol, benzenemethanol,2-chloro-4-fluoro-a-methyl, acmc-1cg87, alphas-2-chloro-4-fluoro-alpha-methylbenzenemethanol
PubChem CID 14501805
IUPAC Name 1-(2-chloro-4-fluorophenyl)ethanol
SMILES CC(O)C1=C(Cl)C=C(F)C=C1

Specifications

Refractive Index 1.521
Quantity 5 g
Formula Weight 174.6
Percent Purity 95%
Chemical Name or Material 1-(2-Chloro-4-fluorophenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.