missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)-2-pyrrolidone, 95%, Thermo Scientific Chemicals

Catalog Number p-7077433
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
51333-90-5
C6H10ClNO
147.60
MFCD00169064
CWLYHDWHNFLUEI-UHFFFAOYSA-N
1-2-chloroethyl pyrrolidin-2-one, 1-2-chloro-ethyl-pyrrolidin-2-one, 1-2-chloroethyl-2-pyrrolidinone, 2-pyrrolidinone, 1-2-chloroethyl, n-2-chloroethyl-2-pyrrolidone, 1-2-chloroethyl-2-pyrrolidone, n-beta-chloroethyl-pyrrolidinone, 1-2-chloroethyl-pyrrolidin-2-one, n-beta-chloroethyl-2-pyrrolidinone
3156650
1-(2-chloroethyl)pyrrolidin-2-one
ClCCN1CCCC1=O
This item is not returnable. View return policy
51333-90-5
C6H10ClNO
147.60
MFCD00169064
CWLYHDWHNFLUEI-UHFFFAOYSA-N
1-2-chloroethyl pyrrolidin-2-one, 1-2-chloro-ethyl-pyrrolidin-2-one, 1-2-chloroethyl-2-pyrrolidinone, 2-pyrrolidinone, 1-2-chloroethyl, n-2-chloroethyl-2-pyrrolidone, 1-2-chloroethyl-2-pyrrolidone, n-beta-chloroethyl-pyrrolidinone, 1-2-chloroethyl-pyrrolidin-2-one, n-beta-chloroethyl-2-pyrrolidinone
3156650
1-(2-chloroethyl)pyrrolidin-2-one
ClCCN1CCCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 147.61
Percent Purity 95%
Chemical Name or Material 1-(2-Chloroethyl)-2-pyrrolidone
compliance-icons
Product Identifier
  • 1-(2-Chloroethyl)-2-pyrrolidone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'