missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, TRC

Product Code. 30386837
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386837 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386837 Supplier Toronto Research Chemicals Supplier No. TRCD46061525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol
CAS 2190-15-0
Purity Grade Notes HPLC
Molecular Formula C55 H98 O6
Quantity 25 mg
Synonym PLL, 9,12-Octadecadienoic acid (9Z,12Z)-, 1,1'-[1-[[(1-oxohexadecyl)oxy]methyl]-1,2-ethanediyl] ester, 9,12-Octadecadienoic acid (9Z,12Z)-, 1-[[(1-oxohexadecyl)oxy]methyl]-1,2-ethanediyl ester (9CI), 9,12-Octadecadienoic acid (Z,Z)-, 1-[[(1-oxohexadecyl)oxy]methyl]-1,2-ethanediyl ester, Linolein, 3-palmito-1,2-di- (7CI,8CI), Palmitin, 2,3-dilinoleo-1- (8CI), 1,2-Dilinoleoyl-3-palmitin, 1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, 1-Palmito-2,3-dilinolein
InChI Formula InChI=1S/C55H98O6/c1-4-7-10-13-16-19-22-25-27-30-33-36-39-42-45-48-54(57)60-51-52(50-59-53(56)47-44-41-38-35-32-29-24-21-18-15-12-9-6-3)61-55(58)49-46-43-40-37-34-31-28-26-23-20-17-14-11-8-5-2/h16-17,19-20,25-28,52H,4-15,18,21-24,29-51H2,1-3H3/b19-16-,20-17-,27-25-,28-26-
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/C\C=C/CCCCC)OC(=O)CCCCCCC\C=C/C\C=C/CCCCC
IUPAC Name [3-hexadecanoyloxy-2-[(9Z,12Z)-octadeca-9,12-dienoyl]oxypropyl] (9Z,12Z)-octadeca-9,12-dienoate
Molecular Weight (g/mol) 855.36
Formula Weight 854.7363
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.