missing translation for 'onlineSavingsMsg'
Learn More

1,2-Ethanedithiol, 98+%

Product Code. p-7046032
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11353416

Brand: Thermo Scientific Alfa Aesar L12865.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 540-63-6 | C2H6S2 | 94.19 g/mol

1,2-Ethanedithiol is used as a reagent in organic synthesis to convert carbonyl compounds to thioacetals (1,3-dithiolanes). It replaces the toxic reagent arsenic tirchloride, which is involved in the synthesis of membrane-permeant fluorogenic biarsenicals from precursor dyes fluorescein and resorufin. It acts bidendate ligand to form complexes with metal ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Ethanedithiol is used as a reagent in organic synthesis to convert carbonyl compounds to thioacetals (1,3-dithiolanes). It replaces the toxic reagent arsenic tirchloride, which is involved in the synthesis of membrane-permeant fluorogenic biarsenicals from precursor dyes fluorescein and resorufin. It acts bidendate ligand to form complexes with metal ions.

Solubility
Miscible with ethanol, acetone, ether and benzene. Slightly miscible with water.

Notes
Air sensitive. Keep away from sources of ignition. Incompatible with alkali metals. Avoid inhalation of its vapor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-63-6
Molecular Formula C2H6S2
Molecular Weight (g/mol) 94.19
MDL Number MFCD00004892
InChI Key VYMPLPIFKRHAAC-UHFFFAOYSA-N
Synonym 1,2-ethanedithiol, 1,2-dimercaptoethane, dithioglycol, ethylene mercaptan, ethylenedithiol, 1,2-ethanethiol, ethylenedimercaptan, dithioethyleneglycol, ethylene dithioglycol, s-ethylene dimercaptan
PubChem CID 10902
IUPAC Name ethane-1,2-dithiol
SMILES C(CS)S

Specifications

Melting Point -41°C
Density 1.123
Boiling Point 144°C to 146°C
Flash Point 45°C (113°F)
Odor Stench
Linear Formula HSCH2CH2SH
Refractive Index 1.558
Quantity 25 g
UN Number UN3071
Beilstein 505946
Sensitivity Air Sensitive
Merck Index 14,3725
Solubility Information Miscible with ethanol,acetone,ether and benzene. Slightly miscible with water.
Formula Weight 94.2
Percent Purity ≥98%
Chemical Name or Material 1,2-Ethanedithiol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Ethanedithiol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Ethanedithiol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.