Learn More
1-[2-(Ethylsulfonyl)ethyl]-2-methyl-4-nitro-1H-imidazole, Mikromol™
Description
1-[2-(Ethylsulfonyl)ethyl]-2-methyl-4-nitro-1H-imidazole
Specifications
Specifications
| Chemical Name or Material | 1-[2-(ethylsulfonyl)ethyl]-2-methyl-4-nitro-1H-imidazole |
| CAS | 25459-12-5 |
| Analyte or Component Names | 1-[2-(ethylsulfonyl)ethyl]-2-methyl-4-nitro-1H-imidazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H13 N3 O4 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Tinidazole Imp. B (EP), Tinidazole USP Related Compound B, Tinidazole USP RC B, 1-[2-(Ethylsulfonyl)ethyl]-2-methyl-4-nitro-1H-imidazole, 1-[2-(Ethylsulfonyl)ethyl]-2-methyl-4-nitro-imidazole, Tinidazole RC B (USP), Tinidazole Related Compound B, Tinidazole Impurity B |
| InChI Formula | InChI=1S/C8H13N3O4S/c1-3-16(14,15)5-4-10-6-8(11(12)13)9-7(10)2/h6H,3-5H2,1-2H3 |
| SMILES | CCS(=O)(=O)CCn1cc(nc1C)[N+](=O)[O-] |
| IUPAC Name | 1-(2-ethylsulfonylethyl)-2-methyl-4-nitroimidazole |
| Show More |