missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluoro-6-nitrophenyl)piperidine, 97%, Thermo Scientific™

Catalog Number 15400189
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1233952-03-8
C10H11FN2O2
210.21
MFCD13689280
QJQZIIGCNJGTMH-UHFFFAOYSA-N
1-2-fluoro-6-nitrophenyl pyrrolidine
53255583
1-(2-fluoro-6-nitrophenyl)pyrrolidine
[O-][N+](=O)C1=C(N2CCCC2)C(F)=CC=C1
This item is not returnable. View return policy
1233952-03-8
C10H11FN2O2
210.21
MFCD13689280
QJQZIIGCNJGTMH-UHFFFAOYSA-N
1-2-fluoro-6-nitrophenyl pyrrolidine
53255583
1-(2-fluoro-6-nitrophenyl)pyrrolidine
[O-][N+](=O)C1=C(N2CCCC2)C(F)=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Formula Weight 224.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2-Fluoro-6-nitrophenyl)piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'