Learn More
1-(2-Fluorophenyl)-1,2,3,4-tetrahydroisoquinoline, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(2-Fluorophenyl)-1,2,3,4-tetrahydroisoquinoline |
| CAS | 1017125-85-7 |
| Molecular Formula | C15H14NF |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C15H14FN/c16-14-8-4-3-7-13(14)15-12-6-2-1-5-11(12)9-10-17-15/h1-8,15,17H,9-10H2 |
| SMILES | Fc1ccccc1C2NCCc3ccccc23 |
| IUPAC Name | 1-(2-fluorophenyl)-1,2,3,4-tetrahydroisoquinoline |
| Molecular Weight (g/mol) | 227.28 |