Learn More
1,2-Hexanediol, TRC
Product Code. 30405452
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405452 10 g
30405453 100 g
30391414 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30405452 Supplier Toronto Research Chemicals Supplier No. TRCH29424510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6920-22-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.17
Synonym (±)-Hexane-1,2-diol, 1,2-Dihydroxyhexane, 1,2-Hexyleneglycol, 5,6-Dihydroxyhexane, DL-1,2-Hexanediol, KMO 6
IUPAC Name hexane-1,2-diol
SMILES CCCCC(O)CO
Specifications
Quantity 10 g
InChI Formula InChI=1S/C6H14O2/c1-2-3-4-6(8)5-7/h6-8H,2-5H2,1H3
Formula Weight 118.0994
Chemical Name or Material 1,2-Hexanediol