Learn More
1-(2-Phenylethyl)piperazine, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(2-Phenylethyl)piperazine |
| CAS | 5321-49-3 |
| Molecular Formula | C32 H58 O5 Si |
| Quantity | 5 g |
| Synonym | 1-Phenethylpiperazine, 4-Phenethylpiperazine, N-(2-Phenylethyl)piperazine, N-(β-Phenylethyl)piperazine, N-Phenethylpiperazine |
| InChI Formula | InChI=1S/C32H58O5Si/c1-21(10-9-15-33)23-11-12-24-28-25(19-27(34)31(23,24)6)30(5)13-14-32(35-16-17-36-32)20-22(30)18-26(28)37-38(7,8)29(2,3)4/h21-28,33-34H,9-20H2,1-8H3/t21?,22-,23?,24-,25?,26+,27-,28?,30-,31+/m0/s1 |
| SMILES | C=1C=CC(=CC1)CCN2CCNCC2 |
| IUPAC Name | (5S,7R,10S,12S,13R,14S)-7-[tert-butyl(dimethyl)silyl]oxy-17-(5-hydroxypentan-2-yl)-10,13-dimethylspiro[1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthrene-3,2'-1,3-dioxolane]-12-ol |
| Molecular Weight (g/mol) | 550.885 |
| Formula Weight | 550.405 |
Product Title
¿Detecta una oportunidad de mejora?