missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyrimidinyl)piperazine dihydrochloride, 98+%, Thermo Scientific™

Product Code. 11431087
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11431087

Brand: Thermo Scientific Alfa Aesar B24085.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94021-22-4
Molecular Formula C8H14Cl2N4
Molecular Weight (g/mol) 237.128
MDL Number MFCD00012779
InChI Key ZNZGJSLHXOMREP-UHFFFAOYSA-N
Synonym 2-1-piperazinyl pyrimidine dihydrochloride, 1-2-pyrimidyl piperazine dihydrochloride, 2-piperazin-1-yl pyrimidine dihydrochloride, 1-2-pyrimidinyl piperazine dihydrochloride, 2-piperazinylpyrimidine hcl, 2-piperazinylpyrimidine dihydrochloride, 2-piperazin-1-ylpyrimidine dihydrochloride, pyrimidine, 2-1-piperazinyl-, dihydrochloride, pyrimidinylpiperazine dihydrochloride, pubchem7349
PubChem CID 11680272
IUPAC Name 2-piperazin-1-ylpyrimidine;dihydrochloride
SMILES C1CN(CCN1)C2=NC=CC=N2.Cl.Cl

Specifications

Melting Point ∼280°C (decomposition)
Quantity 25 g
Beilstein 151178
Sensitivity Hygroscopic
Formula Weight 237.13
Percent Purity ≥98%
Chemical Name or Material 1-(2-Pyrimidinyl)piperazine dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.