missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedithiol, 95%, Thermo Scientific Chemicals

Catalog Number p-7077602
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
626-04-0
C6H6S2
142.23
MFCD00004842
ZWOASCVFHSYHOB-UHFFFAOYSA-N
1,3-benzenedithiol, 1,3-dimercaptobenzene, dithioresorcin, dithioresorcinol, 1,3-benzedithiol, acmc-20ap5g, ksc357g0j
522062
benzene-1,3-dithiol
SC1=CC(S)=CC=C1
This item is not returnable. View return policy
626-04-0
C6H6S2
142.23
MFCD00004842
ZWOASCVFHSYHOB-UHFFFAOYSA-N
1,3-benzenedithiol, 1,3-dimercaptobenzene, dithioresorcin, dithioresorcinol, 1,3-benzedithiol, acmc-20ap5g, ksc357g0j
522062
benzene-1,3-dithiol
SC1=CC(S)=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 24.0°C
Density 1.2400g/mL
Boiling Point 128°C to 129°C (21.0 hPa)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula C6H4(SH)2
Quantity 1 g
Specific Gravity 1.24
Formula Weight 142.24
Percent Purity 95%
Chemical Name or Material 1,3-Benzenedithiol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Benzenedithiol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'