Learn More
1,3-Bis[2,6-bis(1-methylethyl)phenyl]-2-carboxy-1H-imidazolium Inner Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 1,3-Bis[2,6-bis(1-methylethyl)phenyl]-2-carboxy-1H-imidazolium Inner Salt |
| CAS | 917604-39-8 |
| Molecular Formula | C28H36N2O2 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C28H36N2O2/c1-17(2)21-11-9-12-22(18(3)4)25(21)29-15-16-30(27(29)28(31)32)26-23(19(5)6)13-10-14-24(26)20(7)8/h9-20H,1-8H3 |
| SMILES | O=C(C1=[N+](C2=C(C(C)C)C=CC=C2C(C)C)C=CN1C3=C(C(C)C)C=CC=C3C(C)C)[O-] |
| IUPAC Name | 1,3-bis[2,6-di(propan-2-yl)phenyl]imidazol-1-ium-2-carboxylate |
| Molecular Weight (g/mol) | 432.6 |
| Formula Weight | 432.2777 |