missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis[3-[4-(5-chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)piperidin-1-yl]propyl]-1,3-dihydro-2H-benzimidazol-2-one, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
1,3-Bis[3-[4-(5-chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)piperidin-1-yl]propyl]-1,3-dihydro-2H-benzimidazol-2-one
Specifications
Specifications
| Chemical Name or Material | 1,3-Bis[3-[4-(5-chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)piperidin-1-yl]propyl]-1,3-dihydro-2H-benzimidazol-2-one |
| CAS | 1391053-55-6 |
| Analyte or Component Names | 1,3-Bis[3-[4-(5-chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)piperidin-1-yl]propyl]-1,3-dihydro-2H-benzimidazol-2-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C37 H42 Cl2 N8 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Domperidone Imp. F (EP), 1,3-Bis[3-[4-(5-chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)piperidin-1-yl]propyl]-1,3-dihydro-2H-benzimidazol-2-one, Domperidone Impurity F |
| InChI Formula | InChI=1S/C13H19Cl/c1-9-6-11(13(3,4)5)7-10(2)12(9)8-14/h6-7H,8H2,1-5H6 |
| SMILES | Clc1ccc2N(C3CCN(CCCN4C(=O)N(CCCN5CCC(CC5)N6C(=O)Nc7cc(Cl)ccc67)c8ccccc48)CC3)C(=O)Nc2c1 |
| IUPAC Name | 1,3-bis[3-[4-(5-chloro-2-oxo-3H-benzimidazol-1-yl)piperidin-1-yl]propyl]benzimidazol-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction