Learn More
1,3-Cyclohexadiene, 96%, stab. with 0.1% BHT
Product Code. p-7024546
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11403997
Brand: Thermo Scientific Alfa Aesar A19394.06
Request Bulk or Custom Format
CAS: 592-57-4 | C6H8 | 80.13 g/mol
1,3-Cyclohexadiene is used as a hydrogen donor in transfer hydrogenation. It is used in the conversion to benzene. It is useful in the study of proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Cyclohexadiene is used as a hydrogen donor in transfer hydrogenation. It is used in the conversion to benzene. It is useful in the study of proteomics research.
Solubility
Slightly miscible with methanol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 592-57-4
Molecular Formula C6H8
Molecular Weight (g/mol) 80.13
MDL Number MFCD00001532
InChI Key MGNZXYYWBUKAII-UHFFFAOYSA-N
Synonym 1,3-cyclohexadiene, cyclohexadiene, 1,2-dihydrobenzene, unii-jv5w0eg5bp, jv5w0eg5bp, 3-cyclohexen-1,2-ylene, 1,3-cyclohexadien, 1,3 cyclohexadiene, 2,4-cyclohexadiene, cyclohexadiene-1,3
PubChem CID 11605
ChEBI CHEBI:37610
IUPAC Name cyclohexa-1,3-diene
SMILES C1CC=CC=C1
Specifications
Melting Point -89°C
Density 0.847
Boiling Point 79°C to 81°C
Flash Point −8°C (−144°F)
Odor Unpleasant
Refractive Index 1.474
Quantity 5 g
UN Number UN3295
Beilstein 506024
Solubility Information Slightly miscible with methanol. Immiscible with water.
Formula Weight 80.13
Percent Purity 96%
Chemical Name or Material 1,3-Cyclohexadiene, stabilized with 0.1% BHT
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Cyclohexadiene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : 1,3-Cyclohexadiene