missing translation for 'onlineSavingsMsg'
Learn More

1,3-Di-tert-butylbenzimidazolium chloride, 96%, ACROS Organics™

Brand:  Acros Organics 435080010

Additional Details : CAS : 946607-10-9 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11986651

  • € 123.00 / 1g


In stock, ready to ship: 1
Add to basket

Description

Description

Specifications

Chemical Identifiers

946607-10-9
266.81
1,3-di-tert-butylbenzimidazolium chloride, 1,3-di-tert-butyl-1,3-benzodiazol-1-ium chloride, 1,3-di-tert-butyl-1h-benzimidazol-3-ium chloride, 1,3-di-tert-butyl-1h-benzo d imidazol-3-ium chloride
1,3-ditert-butylbenzimidazol-3-ium;chloride
C15H23ClN2
CMQDFXSMZLANNA-UHFFFAOYSA-M
16218265
CC(C)(C)N1C=[N+](C2=CC=CC=C21)C(C)(C)C.[Cl-]

Specifications

1, 3-Di-tert-butylbenzimidazolium chloride
Authentic
Tan to White
1g
95.0
96%
1,3-di-tert-butylbenzimidazolium chloride, 1,3-di-tert-butyl-1,3-benzodiazol-1-ium chloride, 1,3-di-tert-butyl-1h-benzimidazol-3-ium chloride, 1,3-di-tert-butyl-1h-benzo d imidazol-3-ium chloride
CC(C)(C)N1C=[N+](C2=CC=CC=C21)C(C)(C)C.[Cl-]
266.81
266.81
96%
64.15 to 70.90% C, 9.97 to 11.02% N
Glass Bottle
204.0°C
946607-10-9
100.0
C15H23ClN2
CMQDFXSMZLANNA-UHFFFAOYSA-M
1,3-ditert-butylbenzimidazol-3-ium;chloride
16218265
Powder or Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,3-Di-tert-butylbenzimidazolium chloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates