missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diaminoacetone Dihydrochloride Hydrate, TRC

Product Code. 30132834
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132834 1 g 1g
30138046 500 mg 500mg
30007677 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132834 Supplier Toronto Research Chemicals Supplier No. TRCD4161481G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C3 H8 N2 O . 2[H Cl] . xH2 O
Molecular Weight (g/mol) 179.046
IUPAC Name 1,3-diaminopropan-2-one;hydrate;dihydrochloride
SMILES O=C(CN)CN.Cl.Cl.[xH2O]

Specifications

Analyte or Component Names 1,3 Diaminoacetone Dihydrochloride monohydrate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C3H8N2O.2ClH.H2O/c4-1-3(6)2-5;;;/h1-2,4-5H2;2*1H;1H2
Solution Type Neat
Industry Type Pharmaceutical
Quantity 1 g
Formula Weight 178.0276
Chemical Name or Material 1,3 Diaminoacetone Dihydrochloride monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.