missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dibromo-5,5-dimethylhydantoin, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 187.0°C to 191.0°C |
Color | Cream or Yellow |
Flash Point | 155°C |
Loss on Drying | 0.5% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (Iodometry) |
Packaging | Plastic bottle |
Quantity | 25 g |
Show More |
Safety and Handling
Product Identifier
- 1,3-Dibromo-5,5-dimethylhydantoin
- Danger
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'