missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-(trifluoromethoxy)benzene, 97%, Thermo Scientific™

Catalog Number p-7055514
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
207226-31-1
C7H3Br2F3O
319.90
MFCD00075280
UKHOUWWEEAOCTI-UHFFFAOYSA-N
1,3-dibromo-5-trifluoromethoxy benzene, 1,3-dibromo-5-trifluoromethoxy-benzene, 3,5-dibromotrifluoromethoxybenzene, 3,5-dibromo trifluoromethoxy benzene, 3,5-dibromo-1-trifluoromethoxy benzene, 3,5-dibromo-alpha,alpha,alpha-trifluoroanisole, acmc-1cfab, ksc496c1r, 3,5-dibromo-triffluoromethoxybenzene, 3,5-dibromo-, ,-trifluoroanisole
2761179
1,3-dibromo-5-(trifluoromethoxy)benzene
FC(F)(F)OC1=CC(Br)=CC(Br)=C1
This item is not returnable. View return policy
207226-31-1
C7H3Br2F3O
319.90
MFCD00075280
UKHOUWWEEAOCTI-UHFFFAOYSA-N
1,3-dibromo-5-trifluoromethoxy benzene, 1,3-dibromo-5-trifluoromethoxy-benzene, 3,5-dibromotrifluoromethoxybenzene, 3,5-dibromo trifluoromethoxy benzene, 3,5-dibromo-1-trifluoromethoxy benzene, 3,5-dibromo-alpha,alpha,alpha-trifluoroanisole, acmc-1cfab, ksc496c1r, 3,5-dibromo-triffluoromethoxybenzene, 3,5-dibromo-, ,-trifluoroanisole
2761179
1,3-dibromo-5-(trifluoromethoxy)benzene
FC(F)(F)OC1=CC(Br)=CC(Br)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Boiling Point 83°C to 85°C (20 mmHg)(literature)
Flash Point >113°C (235°F)
Refractive Index 1.508
Quantity 5 g
Formula Weight 319.9
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1,3-Dibromo-5-(trifluoromethoxy)benzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'