missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloroacetone Ethylene Ketal, TRC

Product Code. 30378810
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378810 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378810 Supplier Toronto Research Chemicals Supplier No. TRCD431780500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dichloroacetone Ethylene Ketal
CAS 26271-50-1
Molecular Formula C5H8Cl2O2
Quantity 500 mg
Synonym 2,2-Bis(chloromethyl)-1,3-dioxolane, NSC 131448
InChI Formula InChI=1S/C5H8Cl2O2/c6-3-5(4-7)8-1-2-9-5/h1-4H2
SMILES ClCC1(CCl)OCCO1
IUPAC Name 2,2-bis(chloromethyl)-1,3-dioxolane
Molecular Weight (g/mol) 171.02
Formula Weight 169.9901
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.