missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodopropane, 99%, stabilized, Thermo Scientific Chemicals

Catalog Number p-113168
61.40 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
627-31-6
C3H6I2
295.88
MFCD00001097
AAAXMNYUNVCMCJ-UHFFFAOYSA-N
trimethylene diiodide, propane, 1,3-diiodo, trimethylene iodide, propane,3-diiodo, pubchem3858, 1,3-bis iodanyl propane, acmc-1av5p, 4-01-00-00228 beilstein handbook reference, 1,3-diiodopropane, contains copper as stabilizer
12314
1,3-diiodopropane
C(CI)CI
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
627-31-6
C3H6I2
295.88
MFCD00001097
AAAXMNYUNVCMCJ-UHFFFAOYSA-N
trimethylene diiodide, propane, 1,3-diiodo, trimethylene iodide, propane,3-diiodo, pubchem3858, 1,3-bis iodanyl propane, acmc-1av5p, 4-01-00-00228 beilstein handbook reference, 1,3-diiodopropane, contains copper as stabilizer
12314
1,3-diiodopropane
C(CI)CI

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Pink to Yellow
Density 2.5760g/mL
Boiling Point 111.0°C to 113.0°C (31.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula I(CH2)3I
Refractive Index 1.6405 to 1.6425
Quantity 25 mL
Beilstein 01, 115
Specific Gravity 2.576
Solubility Information Solubility in water: insoluble in water.
Stabilizer copper
Formula Weight 295.88
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,3-Diiodopropane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Diiodopropane, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'