Learn More
1,3-Dimethyl-6-methylamino-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxaldehyde, 96%, Thermo Scientific™
Code produit 15435135
Modifier l'affichage
Click to view available options
Quantity:
500 mg
Conditionnement:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
15435135 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15435135 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur H31650.ME
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Spécification
CAS 89549-92-8
Melting Point 202°C to 204°C
Molecular Formula C8H11N3O3
MDL Number MFCD10007818
Quantity 500 mg
Sensitivity Air Sensitive
Synonym 1,3-dimethyl-6-methylamino-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde, 1,3-dimethyl-6-methylamino-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxaldehyde, 1,3-dimethyl-4-methylamino-2,6-dioxopyrimidine-5-carbaldehyde
InChI Key LMOMYYSBGBBWCF-UHFFFAOYSA-N
SMILES CNC1=C(C(=O)N(C(=O)N1C)C)C=O
IUPAC Name 1,3-dimethyl-4-(methylamino)-2,6-dioxopyrimidine-5-carbaldehyde
Molecular Weight (g/mol) 197.194
PubChem CID 13223304
Formula Weight 197.2
Percent Purity 96%
Chemical Name or Material 1,3-Dimethyl-6-methylamino-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxaldehyde