Learn More
1-(3-Fluorophenyl)ethanone, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 455-36-7 |
| Molecular Formula | C8H7FO |
| MDL Number | MFCD00000338 |
| Quantity | 100 g |
| SMILES | CC(C1=CC=CC(F)=C1)=O |
| Molecular Weight (g/mol) | 138.14 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-(3-Fluorophenyl)ethanone |