Learn More
1-(3-Fluorophenyl)pyrrole, 98%, Thermo Scientific™
Product Code. 15476157
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476157 1 g 1g
15486157 5 g 5g
15496157 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15476157 Supplier Thermo Scientific Alfa Aesar Supplier No. H33348.03
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
CAS 53871-27-5
Molecular Formula C10H8FN
Molecular Weight (g/mol) 161.18
MDL Number MFCD02665249
InChI Key MMILNBLSOHVLCJ-UHFFFAOYSA-N
Synonym 1-3-fluorophenyl pyrrole, 1-3-fluorophenyl-1h-pyrrole
PubChem CID 4763720
IUPAC Name 1-(3-fluorophenyl)pyrrole
SMILES FC1=CC=CC(=C1)N1C=CC=C1
Specifications
Boiling Point 54°C (0.15 mmHg)
Refractive Index 1.578
Quantity 1 g
Formula Weight 161.18
Percent Purity 98%
Chemical Name or Material 1-(3-Fluorophenyl)pyrrole