Learn More
1,3-Propanediol di-p-toluenesulfonate, 98%, Thermo Scientific Chemicals
Description
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as pharmaceutical intermediate.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | 90°C to 92°C |
| Quantity | 5 g |
| Formula Weight | 384.47 |
| Percent Purity | 98% |
| Chemical Name or Material | 1,3-Propanediol di-p-toluenesulfonate |