Learn More
1-(4-Aminobutyl)guanidine sulfate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2482-00-0 |
| Molecular Formula | C5H16N4O4S |
| MDL Number | MFCD00013109 |
| Quantity | 100 g |
| SMILES | NC(NCCCCN)=N.O=S(O)(O)=O |
| Molecular Weight (g/mol) | 228.27 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-(4-Aminobutyl)guanidine sulfate |