Learn More
1,4-Bis(2-methylstyryl)benzene, 99%
Product Code. 11475643
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11475643
Brand: Thermo Scientific Alfa Aesar A13534.06
Request Bulk or Custom Format
CAS: 13280-61-0 | C24H22 | 310.44 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13280-61-0
Molecular Formula C24H22
Molecular Weight (g/mol) 310.44
MDL Number MFCD00008529
InChI Key QKLPIYTUUFFRLV-YTEMWHBBSA-N
Synonym 1,4-bis 2-methylstyryl benzene, bis-msb, p-bis o-methylstyryl benzene, benzene, 1,4-bis 2-2-methylphenyl ethenyl, 1,4-di 2-methylstyryl benzene, 1,4-bis 2-methylstyryl benzene bis-msb, benzene, p-bis o-methylstyryl, 1,4-di-2-methylstyryl benzene, 2-menthylstyrene, 1-methyl-2-2-4-2-2-methylphenyl ethenyl phenyl ethenyl benzene
PubChem CID 5378735
IUPAC Name 1,4-bis[(E)-2-(2-methylphenyl)ethenyl]benzene
SMILES CC1=CC=CC=C1C=CC2=CC=C(C=C2)C=CC3=CC=CC=C3C
Specifications
Melting Point 179°C to 184°C
Odor Odorless
Quantity 5 g
Beilstein 2375845
Sensitivity Light sensitive
Formula Weight 310.44
Percent Purity 99%
Chemical Name or Material 1,4-Bis(2-methylstyryl)benzene