Learn More
1-(4-Bromophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%, Alfa Aesar™
Brand:  Alfa Aesar H65396.MD
Additional Details : CAS : 587-40-6 Weight : 0.00025kg
Quantity 250mg
 View more versions of this product
Product Code. 15439616
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Chemical Identifiers
587-40-6
393.588
FYKRQZIQTIMSMT-UHFFFAOYSA-N
17383110
C1=CC(=CC=C1NC(=O)NC2=CC(=C(C=C2)Cl)C(F)(F)F)Br
C14H9BrClF3N2O
MFCD06188835
1-4-bromophenyl-3-4-chloro-3-trifluoromethyl phenyl urea, 3-4-bromophenyl-1-4-chloro-3-trifluoromethyl phenyl urea, n-4-bromophenyl 4-chloro-3-trifluoromethyl phenyl amino carboxamide
1-(4-bromophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
Specifications
1-(4-Bromophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
587-40-6
MFCD06188835
FYKRQZIQTIMSMT-UHFFFAOYSA-N
1-(4-bromophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
17383110
Solid
250mg
C14H9BrClF3N2O
1-4-bromophenyl-3-4-chloro-3-trifluoromethyl phenyl urea, 3-4-bromophenyl-1-4-chloro-3-trifluoromethyl phenyl urea, n-4-bromophenyl 4-chloro-3-trifluoromethyl phenyl amino carboxamide
C1=CC(=CC=C1NC(=O)NC2=CC(=C(C=C2)Cl)C(F)(F)F)Br
393.588
393.59
97%