Learn More
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific™
Catalog Number p-3586572
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
62546-27-4
C11H11BrN2
251.127
MFCD09817558
ATCMNDCOEJOOSF-UHFFFAOYSA-N
1-4-bromophenyl-3,5-dimethyl-1h-pyrazole, 1-4-bromophenyl-3,5-dimethylpyrazole, 1h-pyrazole, 1-4-bromophenyl-3,5-dimethyl, acmc-209n4v
24229761
1-(4-bromophenyl)-3,5-dimethylpyrazole
CC1=CC(=NN1C2=CC=C(C=C2)Br)C
62546-27-4
C11H11BrN2
251.127
MFCD09817558
ATCMNDCOEJOOSF-UHFFFAOYSA-N
1-4-bromophenyl-3,5-dimethyl-1h-pyrazole, 1-4-bromophenyl-3,5-dimethylpyrazole, 1h-pyrazole, 1-4-bromophenyl-3,5-dimethyl, acmc-209n4v
24229761
1-(4-bromophenyl)-3,5-dimethylpyrazole
CC1=CC(=NN1C2=CC=C(C=C2)Br)C
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 251.13
Percent Purity 97%
Chemical Name or Material 1-(4-Bromophenyl)-3, 5-dimethyl-1H-pyrazole