Learn More
1,4-Dibromo-2-fluorobenzene, 98%
Product Code. 11429187
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429187 100 g 100g
11419187 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11429187 Supplier Thermo Scientific Alfa Aesar Supplier No. B21281.22
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 1435-52-5 | C6H3Br2F | 253.896 g/mol
It is used in preparation of 1,4-bis(2-hydroxy-2-methyl-3-butynyl)-2-fluorobenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in preparation of 1,4-bis(2-hydroxy-2-methyl-3-butynyl)-2-fluorobenzene.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 1435-52-5
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.896
MDL Number MFCD00010603
InChI Key WNSNPGHNIJOOPM-UHFFFAOYSA-N
Synonym 2,5-dibromo-fluorobenzene, 2-fluoro-4-bromobromobenzene, 4-bromo-2-fluorobromobenzene, 2,5-dibromofluorobenzene, benzene, 1,4-dibromo-2-fluoro, 1,4-dibromo-2-fluorobenzene, 2,5-dibromo-1-fluorobenzene, pubchem3448, ksc494g9h, 1,4-dibromo-2-fluoro-benzene
PubChem CID 137004
IUPAC Name 1,4-dibromo-2-fluorobenzene
SMILES C1=CC(=C(C=C1Br)F)Br
Specifica
Melting Point 33°C to 35°C
Boiling Point 216°C
Flash Point 101°C (213°F)
Quantity 100 g
Beilstein 3236451
Solubility Information Insoluble in water.
Formula Weight 253.91
Percent Purity 98%
Chemical Name or Material 1,4-Dibromo-2-fluorobenzene
RUO – Research Use Only
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.