missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluorophenyl)-3-(4-methylbenzyl)urea, 97%

Brand:  Alfa Aesar™ H62501.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15439028

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

MFCD03551356

Specifications

1-(4-Fluorophenyl)-3-(4-methylbenzyl)urea
MFCD03551356
Solid
5g
258.29
97%
Videos
Documents