missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Iodophenyl)-1H-pyrazole, ≥95%, Thermo Scientific™

Product Code. p-2694350
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10272632 250 mg Amber Glass Bottle 250mg
10354422 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10272632 Supplier Thermo Scientific Maybridge Supplier No. CC18411CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 368869-86-7
Molecular Formula C9H7IN2
Molecular Weight (g/mol) 270.07
MDL Number MFCD02681941
InChI Key ADIQBEMLKKKISJ-UHFFFAOYSA-N
PubChem CID 2776481
IUPAC Name 1-(4-iodophenyl)pyrazole
SMILES IC1=CC=C(C=C1)N1C=CC=N1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-4-iodophenyl-1h-pyrazole, 1-4-iodophenyl pyrazole, 4-iodophenyl pyrazole, 1-4-iodo-phenyl-1h-pyrazole, 1h-pyrazole,1-4-iodophenyl, 1h-pyrazole, 1-4-iodophenyl
Formula Weight 270.07
Percent Purity 95%
Chemical Name or Material 1-(4-Iodophenyl)-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.