Learn More
1-(4-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(4-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione |
| CAS | 720-94-5 |
| Molecular Formula | C11 H9 F3 O2 |
| Quantity | 1 g |
| Synonym | 1,3-Butanedione','4,4,4-trifluoro-1-(4-methylphenyl)-, 1,3-Butanedione','4,4,4-trifluoro-1-p-tolyl- (6CI,8CI), 4,4,4-Trifluoro-1-(4-methylphenyl)-1,3-butanedione, (4-Methylbenzoyl)trifluoroacetone, 1,1,1-Trifluoro-4-(4-methylphenyl)-2,4-butanedione, 1-(4-Methylphenyl)-4,4,4-trifluorobutane-1,3-dione, 4-(4-Methylphenyl)-1,1,1-trifluorobutane-2,4-dione, 4-Methyl-1-(4,4,4-trifluoro-3-oxobutanoyl)benzene |
| InChI Formula | InChI=1S/C11H9F3O2/c1-7-2-4-8(5-3-7)9(15)6-10(16)11(12,13)14/h2-5H,6H2,1H3 |
| SMILES | Cc1ccc(cc1)C(=O)CC(=O)C(F)(F)F |
| IUPAC Name | 4,4,4-trifluoro-1-(4-methylphenyl)butane-1,3-dione |
| Molecular Weight (g/mol) | 230.18 |
| Formula Weight | 230.0555 |